Order Entry
Czech Republic
ContactUsLinkComponent
Anti-TrpM7 Ion Channel and Ser/Thr Kinase Mouse Monoclonal Antibody
Anti-TrpM7 Ion Channel and Ser/Thr Kinase Mouse Monoclonal Antibody
Catalog # QEDB11519
Supplier:  QED Bioscience
Anti-TrpM7 Ion Channel and Ser/Thr Kinase Mouse Monoclonal Antibody
Catalog # QEDB11519
Supplier:  QED Bioscience

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    TrpM7 Ion Channel and Ser/Thr Kinase
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: TrpM7 Ion Channel and Ser/Thr Kinase
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat