Anti-V5 Tag Mouse Monoclonal Antibody (Biotin)
Catalog # GENSA01735-100
Supplier: Genscript
Specifications
- Antibody type:Tag
- Antigen name:V5 tag
- Antigen symbol:V5
- Clonality:Monoclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Tag
Antigen: V5 Tag (V5 tag)
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: