Order Entry
Czech Republic
ContactUsLinkComponent
Anti-TUBB Mouse Monoclonal Antibody [clone: 2G7D4]
Anti-TUBB Mouse Monoclonal Antibody [clone: 2G7D4]
Catalog # GENSA01717-40
Supplier:  Genscript
undefined
Anti-TUBB Mouse Monoclonal Antibody [clone: 2G7D4]
Catalog # GENSA01717-40
Supplier:  Genscript

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    tubulin, beta class I
  • Antigen symbol:
    TUBB
  • Clonality:
    Monoclonal
  • Clone:
    2G7D4
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Western blot:
    Yes
  • Antigen synonyms:
    tubulin beta polypeptide|tubulin beta-5 chain|M40|beta Ib tubulin|tubulin|beta 5-tubulin|tubulin beta chain|tubulin beta-1 chain|CDCBM6|beta polypeptide|beta-4 tubulin|OK/SW-cl.56|TUBB1|TUBB5
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    40 µg
  • Pk:
    40 µG

Specifications

About this item

Type: Primary
Antigen: TUBB (tubulin, beta class I)
Clonality: monoclonal
Clone: 2G7D4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: