Order Entry
Czech Republic
ContactUsLinkComponent
Anti-B-tag Rabbit Polyclonal Antibody
Anti-B-tag Rabbit Polyclonal Antibody
  GENSA00638-40
 :  Genscript
undefined
Anti-B-tag Rabbit Polyclonal Antibody
  GENSA00638-40
 :  Genscript

 

  • Antibody type:
    Tag
  • Antigen symbol:
    BTAG
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    40 µg
  • Pk:
    40 µG

 

 

Type: Tag
Antigen: B-tag
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: