Anti-MK5 / MAPKAPK5 Goat Polyclonal Antibody
Katalogové číslo EVBIEB11928
Supplier: Everest Biotech
Specifications
- Antibody type:Primary
- Antigen symbol:MK5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:internal
- Cross adsorption:none
- Antigen synonyms:MAPKAPK5
- Amino acid number:192 to 206
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: MK5 / MAPKAPK5
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: