Anti-DNAJC5G Goat Polyclonal Antibody
EVBIEB11850
- Antibody type:Primary
- Antigen name:DnaJ (Hsp40) homolog, subfamily C, member 5 gamma
- Antigen symbol:DNAJC5G
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Epitope:internal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: DNAJC5G (DnaJ (Hsp40) homolog, subfamily C, member 5 gamma)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human