Anti-CRISP2 Goat polyclonal antibody unconjugated
Catalog # EVBIEB09850
Supplier: Everest Biotech
Specifications
- Antibody type:Primary
- Antigen name:Cysteine-Rich Secretory Protein 2
- Antigen symbol:CRISP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Western blot:Yes
- Epitope:internal
- Cross adsorption:none
- Amino acid number:77 to 89
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: CRISP2 (cysteine-rich secretory protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: