Anti-LTK Rabbit Polyclonal Antibody
Catalog # BSBTPA1990
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Leukocyte Receptor Tyrosine Kinase
- Antigen symbol:LTK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:Protein tyrosine kinase-1|TYK1
- Amino acid number:850 to 864
- Sequence:RGLQPQNLWNPTYRS
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human LTK (850-864aa RGLQPQNLWNPTYRS).
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Leukocyte tyrosine kinase receptor(LTK) detection. Tested with WB in Human.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: LTK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human