Anti-Flt3 ligand Rabbit Polyclonal Antibody
Catalog # BSBTPA1914
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Flt3 ligand
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:206 to 220
- Sequence:HWQRTRRRTPRPGEQ
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human Flt3 ligand (206-220aa HWQRTRRRTPRPGEQ).
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Fms-related tyrosine kinase 3 ligand(FLT3LG) detection. Tested with WB in Human.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Flt3 ligand
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human