Anti-RIP2 Rabbit Polyclonal Antibody
Catalog # BSBTPA1861
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Receptor-interacting serine-threonine kinase 2
- Antigen symbol:RIPK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:CARD3|RICK|CARDIAK|receptor interacting serine/threonine kinase 2|GIG30|CCK|RIP2
- Amino acid number:495 to 514
- Sequence:DIQGEEFAKVIVQKLKDNKQ
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human RIP2 (495-514aa DIQGEEFAKVIVQKLKDNKQ), different from the related rat and mouse sequences by one amino acid.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Receptor-interacting serine/threonine-protein kinase 2(RIPK2) detection. Tested with WB in Human; Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: RIP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat