Anti-MCM2 Rabbit Polyclonal Antibody
Catalog # BSBTPA1711
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:DNA replication licensing factor MCM2
- Antigen symbol:MCM2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:MITOTIN|minichromosome maintenance protein 2 homolog|cyclin-like 1|D3S3194|cell devision cycle-like 1|CCNL1|MCM2 minichromosome maintenance deficient 2|mitotin|minichromosome maintenance deficient 2 (mitotin)|DNA replication licensing factor MCM2|nuclear protein BM28|BM28|cdc19|CDCL1
- Amino acid number:884 to 904
- Sequence:DLFKFNKFSRDLKRKLILQQF
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of mouse MCM2 (884-904aa DLFKFNKFSRDLKRKLILQQF), different from the related rat sequence by one amino acid.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for DNA replication licensing factor MCM2(MCM2) detection. Tested with WB, IHC-P, ICC in Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: MCM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat