Order Entry
Czech Republic
ContactUsLinkComponent
Anti-GJB2 Rabbit Polyclonal Antibody
Anti-GJB2 Rabbit Polyclonal Antibody
Catalog # BSBTPA1025
Supplier:  Boster Bio
undefined
Anti-GJB2 Rabbit Polyclonal Antibody
Catalog # BSBTPA1025
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Gap Junction Protein, Beta 2
  • Antigen symbol:
    GJB2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    N-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Antigen synonyms:
    CX26|DFNB1A|DFNB1|NSRD1|HID|DFNA3A|PPK|DFNA3|KID|connexin 26|Gap junction beta-2 protein
  • Amino acid number:
    1 to 16
  • Sequence:
    MDWGTLQTILGGVNKH
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the N-terminus of human GJB2 (1-16aa, MDWGTLQTILGGVNKH), different from the related mouse and rat sequences by two amino acids.
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Rabbit IgG polyclonal antibody for Gap junction beta-2 protein(GJB2) detection. Tested with WB in Human; Mouse; Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: GJB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat