Anti-beta-Actin Mouse Monoclonal Antibody [clone: AC-15]
Catalog # BSBTMA1115
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Actin beta
- Antigen symbol:ACTB
- Clonality:Monoclonal
- Clone:AC-15
- Conjugation:KLH (Keyhole Limpet Hemocyanin)
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Ascites
- Form:Lyophilized
- Antigen synonyms:PS1TP5-binding protein 1|cytoplasmic 1|PS1TP5BP1|BRWS1|beta cytoskeletal actin|actin
- Sequence:Ac-DDDIAALVIDNGSGK
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:slightly modified beta-cytoplasmic actin N-terminal peptide, Ac-Asp-Asp-Asp-Ile-Ala-Ala-Leu-Val-Ile-Asp-Asn-Gly-Ser-Gly-Lys, conjugated to KLH.
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Mouse IgG monoclonal antibody for beta-Actin, actin, beta (ACTB) detection. Tested with WB, IHC-P in Human; mouse; rat. No cross reactivity with other proteins.
Reconstitute with 1,2% sodium acetate or neutral PBS.
If 1 ml of PBS is used, the antibody concentration will be 100 µg/ml.
Type: Primary
Antigen: beta-Actin
Clonality: Monoclonal
Clone: AC-15
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat