Anti-Synaptophysin Mouse Monoclonal Antibody [clone: Syn-58]
BSBTMA1091
- Antibody type:Primary
- Antigen name:Synaptophysin
- Antigen symbol:SYP
- Clonality:Monoclonal
- Clone:Syn-58
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human,Rat
- Western blot:Yes
- Cross adsorption:No
- Format:Ascites
- Form:Lyophilized
- Antigen synonyms:MRXSYP|major synaptic vesicle protein P38|MRX96
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:Rat retina synaptosome.
- Size:100 µg
- Pk:0,1 mg
Mouse IgG monoclonal antibody for Synaptophysin, synaptophysin (SYP) detection. Tested with WB, IHC-F in Human; Rat. No cross reactivity with other proteins.
Reconstitute with 1,2% sodium acetate or neutral PBS.
If 1 ml of PBS is used, the antibody concentration will be 100 µg/ml.
Type: Primary
Antigen: Synaptophysin
Clonality: Monoclonal
Clone: Syn-58
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat