Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-PCK-26 Mouse Monoclonal Antibody [clone: PC-26]
  BSBTMA1082
Anti-PCK-26 Mouse Monoclonal Antibody [clone: PC-26]
  BSBTMA1082

 

  • Antibody type:
    Primary
  • Antigen symbol:
    PCK-26
  • Clonality:
    Monoclonal
  • Clone:
    PC-26
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Ascites
  • Form:
    Lyophilized
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    Cytokeratin from human epidermis.
  • Size:
    100 µg
  • Pk:
    0,1 mg

 

 

Type: Primary
Antigen: PCK-26
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat