Anti-Filensin Mouse Monoclonal Antibody [clone: FIL-27]
Catalog # BSBTMA1041
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Filensin
- Clonality:Monoclonal
- Clone:FIL-27
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human,Bovine
- Western blot:Yes
- Cross adsorption:No
- Format:Ascites
- Form:Lyophilized
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:Human and bovine lens filament enriched fraction (plasma membrane-cytoskeleton complex).
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: Filensin
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Bovine