Order Entry
Czech Republic
ContactUsLinkComponent
Anti-MTNR1B Rabbit Polyclonal Antibody
Anti-MTNR1B Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals
Certificates
undefined
Anti-MTNR1B Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNLS932SS
  • NOVUNLS932
  • Antigen symbol:
  • MTNR1B
  • MTNR1B
  • Antigen name:
  • Melatonin Receptor 1B
  • Melatonin Receptor 1B
  • Antigen synonyms:
  • MT2|FGQTL2|MEL-1B-R
  • MT2|FGQTL2|MEL-1B-R
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 4544
  • 4544
  • Isotype:
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Immunogen:
  • A synthetic peptide made to an internal region of the human Melatonin Receptor 1B protein (within residues 200-300) [Swiss-Prot P49286]
  • A synthetic peptide made to an internal region of the human Melatonin Receptor 1B protein (within residues 200-300) [Swiss-Prot P49286]
  • Purification:
  • Immunogen affinity purified
  • Immunogen affinity purified
  • Concentration:
  • 1 mg/ml
  • 1 mg/ml
  • Storage buffer:
  • PBS and 30% Glycerol
  • PBS and 30% Glycerol
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Tested applications:
  • Store at 4 °C short term. Aliquot and store at –20 °C long term. Avoid freeze-thaw cycles

Specifications

Related Information

Frequently Bought Together