Anti-GPR119 Rabbit Polyclonal Antibody (DyLight® 405)
NOVUNLS548V
- Antibody type:Primary
- Antigen name:G Protein-Coupled Receptor 119
- Antigen symbol:GPR119
- Clonality:Polyclonal
- Conjugation:DyLight® 405
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Mouse
- Gene ID:139760
- Antigen synonyms:GPCR2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 μL
- Pk:50 µl
Type: Primary
Antigen: GPR119 (G protein-coupled receptor 119)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse