Anti-FPR2 Rabbit Polyclonal Antibody (DyLight® 405)
Catalog # NOVUNLS1878V
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Formyl Peptide Receptor 2
- Antigen symbol:FPR2
- Clonality:Polyclonal
- Conjugation:DyLight® 405
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Gene ID:2358
- Antigen synonyms:FPRH1|FPRH2|FMLPX|FPRL1|FPR2A|FMLP-R-II|HM63|LXA4R|ALXR
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 μL
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: FPR2 (formyl peptide receptor 2)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat