Anti-GPR39 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Catalog # NOVUNLS139F
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:G Protein-Coupled Receptor 39
- Antigen symbol:GPR39
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Gene ID:2863
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 μL
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: GPR39 (G protein-coupled receptor 39)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse