Order Entry
Czech Republic
ContactUsLinkComponent
Anti-BNIP3L Rabbit Polyclonal Antibody
Anti-BNIP3L Rabbit Polyclonal Antibody
Dodavatel:  Novus Biologicals
Certifikáty
undefined
Anti-BNIP3L Rabbit Polyclonal Antibody
Dodavatel:  Novus Biologicals

Specifikace

  • Catalog No:
  • NOVUNBP1-78264
  • NOVUNBP1-78264SS
  • Antigen symbol:
  • BNIP3L
  • BNIP3L
  • Antigen name:
  • BCL2 Interacting Protein 3 like
  • BCL2 Interacting Protein 3 like
  • Antigen synonyms:
  • NIP3L|BNIP3A|BNIP3H|BNIP3a|Bnip3L|Adenovirus E1B19K-binding protein B5|NIX
  • NIP3L|BNIP3A|BNIP3H|BNIP3a|Bnip3L|Adenovirus E1B19K-binding protein B5|NIX
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 665
  • 665
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifikace

Související informace

Frequently Bought Together