Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-Lamin A + C R453W Mouse Monoclonal Antibody [clone: 12A-2F5]
 
undefined
Anti-Lamin A + C R453W Mouse Monoclonal Antibody [clone: 12A-2F5]

 

  • Catalog No:
  • NOVUNBP1-77401
  • NOVUNBP1-77401SS
  • Antigen symbol:
  • LMNA
  • LMNA
  • Antigen name:
  • Lamin A/C
  • Lamin A/C
  • Antigen synonyms:
  • LMNA|LMN1|70 kDa lamin|Prelamin-A/C|Lamin-A/C|Renal carcinoma antigen NY-REN-32
  • LMNA|LMN1|70 kDa lamin|Prelamin-A/C|Lamin-A/C|Renal carcinoma antigen NY-REN-32
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 12A-2F5
  • 12A-2F5
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 4000
  • 4000
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

 

 

Frequently Bought Together