Order Entry
Czech Republic
ContactUsLinkComponent
Anti-NFKB1 Rabbit Polyclonal Antibody
Anti-NFKB1 Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals
Certificates
undefined
Anti-NFKB1 Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNBP1-77395
  • NOVUNBP1-77395SS
  • Antigen symbol:
  • NFkB p105/p50
  • NFkB p105/p50
  • Antigen name:
  • Nuclear factor NF-kappa-B p105 subunit
  • Nuclear factor NF-kappa-B p105 subunit
  • Antigen synonyms:
  • EBP-1|Nuclear factor NF-kappa-B p50 subunit|Nuclear factor NF-kappa-B p105 subunit|NFKB1|DNA-binding factor KBF1|Nuclear factor of kappa light polypeptide gene enhancer in B-cells 1
  • EBP-1|Nuclear factor NF-kappa-B p50 subunit|Nuclear factor NF-kappa-B p105 subunit|NFKB1|DNA-binding factor KBF1|Nuclear factor of kappa light polypeptide gene enhancer in B-cells 1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 4790
  • 4790
  • Western blot:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together