Anti-F2R Mouse Monoclonal Antibody (DyLight® 755) [clone: N2-11]
Catalog # NOVUNBP1-71770IR
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:coagulation factor II (thrombin) receptor
- Antigen symbol:F2R
- Clonality:Monoclonal
- Clone:N2-11
- Conjugation:DyLight® 755
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human,Mouse
- Western blot:Yes
- Gene ID:2149
- Antigen synonyms:PAR1|PAR-1|HTR|CF2R|TR
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 Tests
- Pk:100 Tests
Specifications
About this item
Type: Primary
Antigen: F2R (coagulation factor II (thrombin) receptor)
Clonality: monoclonal
Clone: N2-11
Conjugation: DyLight® 755
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse