Order Entry
Czech Republic
ContactUsLinkComponent
Anti-F2R Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: N2-11]
Anti-F2R Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: N2-11]
Catalog # NOVUNBP1-71770F
Supplier:  Novus Biologicals
Anti-F2R Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: N2-11]
Catalog # NOVUNBP1-71770F
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    coagulation factor II (thrombin) receptor
  • Antigen symbol:
    F2R
  • Clonality:
    Monoclonal
  • Clone:
    N2-11
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Gene ID:
    2149
  • Antigen synonyms:
    PAR1|PAR-1|HTR|CF2R|TR
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 μL
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: F2R (coagulation factor II (thrombin) receptor)
Clonality: monoclonal
Clone: N2-11
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse