Order Entry
Czech Republic
ContactUsLinkComponent
Anti-F2R Mouse monoclonal antibody unconjugated [clone: N2-11]
Anti-F2R Mouse monoclonal antibody unconjugated [clone: N2-11]
Supplier:  Novus Biologicals
Certificates
undefined
Anti-F2R Mouse monoclonal antibody unconjugated [clone: N2-11]
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNBP1-71770
  • NOVUNBP1-71770SS
  • Antigen name:
  • Thrombin Receptor
  • Thrombin Receptor
  • Antigen synonyms:
  • PAR1|PAR-1|HTR|CF2R|TR
  • PAR1|PAR-1|HTR|CF2R|TR
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • N2-11
  • N2-11
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 2149
  • 2149
  • Isotype:
  • IgG1
  • IgG1
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together