Order Entry
Czech Republic
ContactUsLinkComponent
Anti-EXOSC9 Rabbit Polyclonal Antibody (DyLight® 650)
Anti-EXOSC9 Rabbit Polyclonal Antibody (DyLight® 650)
Catalog # NOVUNBP1-71702C
Supplier:  Novus Biologicals
undefined
Anti-EXOSC9 Rabbit Polyclonal Antibody (DyLight® 650)
Catalog # NOVUNBP1-71702C
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Exosome Component 9
  • Antigen symbol:
    EXOSC9
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 650
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Chicken,
    Human,
    Mouse
  • Western blot:
    Yes
  • Gene ID:
    5393
  • Antigen synonyms:
    Rrp45p|RRP45|PMSCL1|PM/Scl-75|p6|p5
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µL
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: EXOSC9 (exosome component 9)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Chicken, Human, Mouse