Order Entry
Czech Republic
ContactUsLinkComponent
Anti-DGAT2 Rabbit Polyclonal Antibody (DyLight® 550)
Anti-DGAT2 Rabbit Polyclonal Antibody (DyLight® 550)
Catalog # NOVUNBP1-71701R
Supplier:  Novus Biologicals
Anti-DGAT2 Rabbit Polyclonal Antibody (DyLight® 550)
Catalog # NOVUNBP1-71701R
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Diacylglycerol O-acyltransferase 2
  • Antigen symbol:
    DGAT2
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 550
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Gene ID:
    84649
  • Antigen synonyms:
    HMFN1045|GS1999FULL|ARAT
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µL
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: DGAT2 (diacylglycerol O-acyltransferase 2)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse