Anti-DICER1 Rabbit Polyclonal Antibody
Catalog # NOVUNBP1-71691SS
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Dicer 1, ribonuclease type III
- Antigen symbol:DICER1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Mouse
- Gene ID:23405
- Antigen synonyms:DCR1|MNG1|Dicer|HERNA|RMSE2|K12H4.8-LIKE|Dicer1e
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:25 µL
- Pk:25 µl
Specifications
About this item
The Dicer Antibody from Novus Biologicals is a rabbit polyclonal antibody to Dicer. This antibody reacts with human, mouse. The Dicer Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:50-1:100
Type: Primary
Antigen: Dicer
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse