Anti-PINK1 Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
The PINK1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to PINK1. This antibody reacts with human, mouse. The PINK1 Antibody has been validated for the following applications: Simple Western, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:100
Type: Primary
Antigen: PINK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- NOVUNBP1-49678
- NOVUNBP1-49678SS
- Antigen symbol:
- PINK1
- PINK1
- Antigen name:
- PTEN-Induced Putative Kinase 1
- PTEN-Induced Putative Kinase 1
- Antigen synonyms:
- PARK6|mitochondrial [Precursor]|Serine/threonine kinase PINK1|PTEN induced putative kinase 1|PINK1|BRPK|protein kinase BRPK
- PARK6|mitochondrial [Precursor]|Serine/threonine kinase PINK1|PTEN induced putative kinase 1|PINK1|BRPK|protein kinase BRPK
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 65018
- 65018
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together