Anti-CCR2 Rabbit Polyclonal Antibody (DyLight® 550)
NOVUNBP1-48338R
- Antibody type:Primary
- Antigen name:Chemokine (C-C motif) receptor 2
- Antigen symbol:CCR2
- Clonality:Polyclonal
- Conjugation:DyLight® 550
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Gene ID:729230
- Antigen synonyms:CMKBR2|CKR2|MCP-1-R|CCR-2|CC-CKR-2|CKR2A|CCR2A|CD192|CCR2B|CKR2B
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Type: Primary
Antigen: CCR2 (chemokine (C-C motif) receptor 2)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse