Anti-CCR2 Rabbit polyclonal antibody unconjugated
Supplier: Novus Biologicals
Certificates
About this item
Type: Primary
Antigen: CCR2 (chemokine (C-C motif) receptor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- NOVUNBP1-48338SS
- NOVUNBP1-48338
- Antigen symbol:
- CCR2
- CCR2
- Antigen name:
- Chemokine (C-C motif) receptor 2
- Chemokine (C-C motif) receptor 2
- Antigen synonyms:
- CMKBR2|CKR2|MCP-1-R|CCR-2|CC-CKR-2|CKR2A|CCR2A|CD192|CCR2B|CKR2B
- CMKBR2|CKR2|MCP-1-R|CCR-2|CC-CKR-2|CKR2A|CCR2A|CD192|CCR2B|CKR2B
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 729230
- 729230
- Isotype:
- IgG
- IgG
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together