Anti-PKM Rabbit Polyclonal Antibody (DyLight® 550)
Catalog # NOVUNBP1-48308R
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Pyruvate Kinase, muscle
- Antigen symbol:PKM
- Clonality:Polyclonal
- Conjugation:DyLight® 550
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Gene ID:5315
- Antigen synonyms:THBP1|PKM2|TCB|CTHBP|HEL-S-30|PK3|OIP3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: PKM (pyruvate kinase, muscle)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat