Order Entry
Czech Republic
ContactUsLinkComponent
Anti-HIF1A Rabbit Polyclonal Antibody (DyLight® 650) [clone: exon 13]
Anti-HIF1A Rabbit Polyclonal Antibody (DyLight® 650) [clone: exon 13]
Catalog # NOVUNBP1-47181C
Supplier:  Novus Biologicals
undefined
Anti-HIF1A Rabbit Polyclonal Antibody (DyLight® 650) [clone: exon 13]
Catalog # NOVUNBP1-47181C
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    HIF1 alpha
  • Antigen symbol:
    HIF1A
  • Clonality:
    Polyclonal
  • Clone:
    exon 13
  • Conjugation:
    DyLight® 650
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    3091
  • Antigen synonyms:
    PASD8|HIF-1alpha|HIF1-ALPHA|HIF-1A|HIF1|MOP1|HIF-1-alpha|bHLHe78
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µL
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: HIF1A (hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor))
Clonality: polyclonal
Clone: exon 13
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human