Order Entry
Czech Republic
ContactUsLinkComponent
Anti-HIF1A Rabbit Polyclonal Antibody [clone: exon 13]
Anti-HIF1A Rabbit Polyclonal Antibody [clone: exon 13]
Supplier:  Novus Biologicals
Certificates
Anti-HIF1A Rabbit Polyclonal Antibody [clone: exon 13]
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNBP1-47181SS
  • NOVUNBP1-47181
  • Antigen symbol:
  • HIF1A
  • HIF1A
  • Antigen name:
  • HIF1 alpha
  • HIF1 alpha
  • Antigen synonyms:
  • PASD8|HIF-1alpha|HIF1-ALPHA|HIF-1A|HIF1|MOP1|HIF-1-alpha|bHLHe78
  • PASD8|HIF-1alpha|HIF1-ALPHA|HIF-1A|HIF1|MOP1|HIF-1-alpha|bHLHe78
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • exon 13
  • exon 13
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 3091
  • 3091
  • Western blot:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together