Anti-BMP2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # NOVUNBP1-19751H
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Pregnancy specific beta-1-glycoprotein 4
- Antigen symbol:BMP2
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Gene ID:650
- Antigen synonyms:BMP2A|BDA2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: BMP2 (bone morphogenetic protein 2)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat