Anti-Oval Cell Marker Rat Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: OC2-6E10]
Catalog # NOVUNBP1-18981H
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Oval Cell Marker
- Clonality:Monoclonal
- Clone:OC2-6E10
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rat
- Isotype:IgG1
- Reactivity:Mouse
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: Oval Cell Marker
Clonality: monoclonal
Clone: OC2-6E10
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse