Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Luciferase Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: Luci 21 1-107]
Anti-Luciferase Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: Luci 21 1-107]
Catalog # NOVUNB600-307PE
Supplier:  Novus Biologicals
Anti-Luciferase Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: Luci 21 1-107]
Catalog # NOVUNB600-307PE
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Luciferase
  • Clonality:
    Monoclonal
  • Clone:
    Luci 21 1-107
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 μL
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: Luciferase
Clonality: monoclonal
Clone: Luci 21 1-107
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: