Anti-DIABLO Mouse monoclonal antibody unconjugated [clone: 78-1-118]
Type: Primary
Antigen: DIABLO (diablo, IAP-binding mitochondrial protein)
Clonality: monoclonal
Clone: 78-1-118
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1+IgG3
Reactivity: Human, Mouse, Rat
- Catalog No:
- NOVUNB500-213
- NOVUNB500-213SS
- Antigen symbol:
- SMAC
- SMAC
- Antigen synonyms:
- FLJ10537|SMAC|SMAC3|FLJ25049|DIABLO|DIABLO-S|0610041G12Rik
- FLJ10537|SMAC|SMAC3|FLJ25049|DIABLO|DIABLO-S|0610041G12Rik
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 78-1-118
- 78-1-118
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 56616
- 56616
- Isotype:
- IgG3, IgG1
- IgG3, IgG1
- Western blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Frequently Bought Together