Anti-CHAF1B Mouse Monoclonal Antibody [clone: SS 24 1-68]
Supplier: Novus Biologicals
Certificates
About this item
Type: Primary
Antigen: CHAF1B (chromatin assembly factor 1, subunit B (p60))
Clonality: monoclonal
Clone: SS 24 1-68
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Specifications
- Catalog No:
- NOVUNB500-211
- NOVUNB500-211SS
- Antigen symbol:
- CHAF1B
- CHAF1B
- Antigen name:
- Chromatin Assembly Factor 1, Subunit B (p60)
- Chromatin Assembly Factor 1, Subunit B (p60)
- Antigen synonyms:
- CAF-1|MPHOSPH7|CAF1P60|CAF-IP60|MPP7|CAF1A|CAF1
- CAF-1|MPHOSPH7|CAF1P60|CAF-IP60|MPP7|CAF1A|CAF1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- SS 24 1-68
- SS 24 1-68
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 8208
- 8208
- Isotype:
- IgG1
- IgG1
- Western blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together