Anti-CASP8 Mouse monoclonal antibody unconjugated [clone: FLICE 4-1-20]
Type: Primary
Antigen: CASP8 (caspase 8, apoptosis-related cysteine peptidase)
Clonality: monoclonal
Clone: FLICE 4-1-20
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
- Catalog No:
- NOVUNB500-208SS
- NOVUNB500-208
- Antigen symbol:
- CASP8
- CASP8
- Antigen name:
- Caspase 8
- Caspase 8
- Antigen synonyms:
- FLICE|caspase 8|MACH-beta-1/2/3/4 protein|FADD-homologous ICE/CED-3-like protease|MCH5|apoptotic protease Mch-5|FADD-like ICE|MACH-alpha-1/2/3 protein|CAP4|ALPS2B|apoptotic cysteine protease|Casp-8|MORT1-associated ced-3 homolog|caspase-8|apoptosis-related cysteine protease|ICE-like apoptotic protease 5|MACH
- FLICE|caspase 8|MACH-beta-1/2/3/4 protein|FADD-homologous ICE/CED-3-like protease|MCH5|apoptotic protease Mch-5|FADD-like ICE|MACH-alpha-1/2/3 protein|CAP4|ALPS2B|apoptotic cysteine protease|Casp-8|MORT1-associated ced-3 homolog|caspase-8|apoptosis-related cysteine protease|ICE-like apoptotic protease 5|MACH
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- FLICE 4-1-20
- FLICE 4-1-20
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 841
- 841
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Frequently Bought Together