Order Entry
Czech Republic
ContactUsLinkComponent
Anti-LCAT Rabbit Polyclonal Antibody
Anti-LCAT Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals
Certificates
undefined
Anti-LCAT Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNB400-107SS
  • NOVUNB400-107
  • Antigen symbol:
  • LCAT
  • LCAT
  • Antigen name:
  • Lecithin cholesterol acyltransferase
  • Lecithin cholesterol acyltransferase
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 3931
  • 3931
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together