Order Entry
Czech Republic
ContactUsLinkComponent
Anti-SLIRP Rabbit Polyclonal Antibody
Anti-SLIRP Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals
Certificates
undefined
Anti-SLIRP Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNB110-37258SS
  • NOVUNB110-37258
  • Antigen name:
  • SRA Stem-Loop Interacting RNA Binding Protein
  • SRA Stem-Loop Interacting RNA Binding Protein
  • Antigen synonyms:
  • C14orf156|DC50|PD04872
  • C14orf156|DC50|PD04872
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 81892
  • 81892
  • Western blot:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage buffer:
  • PBS
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together