Anti-KIT Rat Monoclonal Antibody (DyLight® 488) [clone: 2B8]
Catalog # NOVUNB100-77477G
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Proto-oncogene Receptor Tyrosine Kinase
- Antigen symbol:KIT
- Clonality:Monoclonal
- Clone:2B8
- Conjugation:DyLight® 488
- Flow cytometry:Yes
- Host:Rat
- ImmunoPrecipitation:Yes
- Isotype:IgG2b kappa
- Reactivity:Mouse
- Gene ID:3815
- Antigen synonyms:SCFR|CD117|PBT|C-Kit
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: KIT (v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog)
Clonality: monoclonal
Clone: 2B8
Conjugation: DyLight® 488
Epitope:
Host: Rat
Isotype: IgG2b Kappa
Reactivity: Human, Mouse