Order Entry
Czech Republic
ContactUsLinkComponent
Anti-PTPRC Rat Monoclonal Antibody (DyLight® 550) [clone: 30-F11]
Anti-PTPRC Rat Monoclonal Antibody (DyLight® 550) [clone: 30-F11]
  NOVUNB100-77417R
Anti-PTPRC Rat Monoclonal Antibody (DyLight® 550) [clone: 30-F11]
  NOVUNB100-77417R

 

  • Antibody type:
    Primary
  • Antigen name:
    Protein tyrosine phosphatase receptor type C
  • Antigen symbol:
    PTPRC
  • Clonality:
    Monoclonal
  • Clone:
    30-F11
  • Conjugation:
    DyLight® 550
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Mouse
  • Gene ID:
    5788
  • Antigen synonyms:
    CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    0,1 mg

 

 

Type: Primary
Antigen: PTPRC (protein tyrosine phosphatase, receptor type, C)
Clonality: monoclonal
Clone: 30-F11
Conjugation: DyLight® 550
Epitope:
Host: Rat
Isotype: IgG2b Kappa
Reactivity: Mouse