Order Entry
Czech Republic
ContactUsLinkComponent
Anti-CXCR4 Rabbit Polyclonal Antibody
Anti-CXCR4 Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals
Certificates
undefined
Anti-CXCR4 Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNB100-74396SS
  • NOVUNB100-74396
  • Antigen symbol:
  • CXCR4
  • CXCR4
  • Antigen name:
  • Chemokine (C-X-C Motif) Receptor 4
  • Chemokine (C-X-C Motif) Receptor 4
  • Antigen synonyms:
  • HM89|LAP-3|WHIM|NPYR|NPYRL|D2S201E|NPY3R|NPYY3R|LCR1|HSY3RR|CD184|WHIMS|LAP3|LESTR|FB22
  • HM89|LAP-3|WHIM|NPYR|NPYRL|D2S201E|NPY3R|NPYY3R|LCR1|HSY3RR|CD184|WHIMS|LAP3|LESTR|FB22
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 7852
  • 7852
  • Western blot:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage buffer:
  • Whole antisera
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together