Anti-FANCD2 Mouse Monoclonal Antibody [clone: FI-17]
Supplier: Novus Biologicals
Certificates
About this item
Type: Primary
Antigen: FANCD2 (Fanconi anemia, complementation group D2)
Clonality: monoclonal
Clone: FI-17
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Specifications
- Catalog No:
- NOVUNB100-316SS
- NOVUNB100-316
- Antigen symbol:
- FANCD2
- FANCD2
- Antigen name:
- Fanconi Anemia Complementation Group D2
- Fanconi Anemia Complementation Group D2
- Antigen synonyms:
- FAD|FA4|FANCD|FACD|FA-D2|FAD2
- FAD|FA4|FANCD|FACD|FA-D2|FAD2
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- FI-17
- FI-17
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 2177
- 2177
- Western blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ChIP:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together