Anti-HIF-1 beta Mouse Monoclonal Antibody [clone: H1beta234]
Type: Primary
Antigen: HIF-1 beta
Clonality: monoclonal
Clone: H1beta234
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
- Catalog No:
- NOVUNB100-124
- NOVUNB100-124SS
- Antigen symbol:
- HIF1B
- HIF1B
- Antigen name:
- HIF1 beta
- HIF1 beta
- Antigen synonyms:
- class E basic helix-loop-helix protein 2|HIF-1-beta|HIF1BETA|HIF-1beta|dioxin receptor|hypoxia-inducible factor 1|nuclear translocator|bHLHe2|TANGO|HIF1B|beta subunit|HIF1-beta
- class E basic helix-loop-helix protein 2|HIF-1-beta|HIF1BETA|HIF-1beta|dioxin receptor|hypoxia-inducible factor 1|nuclear translocator|bHLHe2|TANGO|HIF1B|beta subunit|HIF1-beta
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- H1beta234
- H1beta234
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 405
- 405
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Gel shift:
- Yes
- Yes
- ChIP:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Frequently Bought Together