Order Entry
Czech Republic
ContactUsLinkComponent
Anti-NUDT1 Rabbit Polyclonal Antibody
Anti-NUDT1 Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals
Certificates
undefined
Anti-NUDT1 Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNB100-109
  • NOVUNB100-109SS
  • Antigen symbol:
  • NUDT1
  • NUDT1
  • Antigen name:
  • 7,8-dihydro-8-oxoguanine triphosphatase
  • 7,8-dihydro-8-oxoguanine triphosphatase
  • Antigen synonyms:
  • NUDIX Motif 1|MTH1|Nucleoside Diphosphate Linked Moiety X Motif 1|NUDT1
  • NUDIX Motif 1|MTH1|Nucleoside Diphosphate Linked Moiety X Motif 1|NUDT1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 4521
  • 4521
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together