Order Entry
Czech Republic
ContactUsLinkComponent
Anti-SCF Rabbit Polyclonal Antibody
Anti-SCF Rabbit Polyclonal Antibody
Catalog # BSBTRP1010
Supplier:  Boster Bio
Anti-SCF Rabbit Polyclonal Antibody
Catalog # BSBTRP1010
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Stem Cell Factor
  • Antigen symbol:
    SCF
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Antigen synonyms:
    c-Kit ligand|SCF|KL-1|kit ligand|familial progressive hyperpigmentation 2|stem cell factor|Kitl|mast cell growth factor|steel factor|SF|FPH2|MGF|DKFZp686F2250|SHEP7
  • Amino acid number:
    26 - 190
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    E. coli-derived human SCF recombinant protein (Position: E26-A190)
  • Purification:
    Purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Rabbit IgG polyclonal antibody for Kit ligand/Mast cell growth factor(KITLG) detection. Tested with WB, ELISA in Human.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: SCF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human